2,3,4-Trifluorotoluene ≥97.0% (by GC)

Supplier: TCI America

T2735-1G T2735-5G
TCT2735-1GEA 76.4 USD
TCT2735-1G TCT2735-5G
2,3,4-Trifluorotoluene ≥97.0% (by GC)
2,3,4-Trifluorotoluene

CAS Number: 193533-92-5
MDL Number: MFCD00075194
Molecular Formula: C7H5F3
Molecular Weight: 146.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 127
Flash Point (°C): 28
Specific Gravity (20/20): 1.24

Formula: C₇H₅F₃
Boiling Pt: 127 °C
Density: 1.24 (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00075194
CAS Number: 193533-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e30a0c577435467dc1a83c9b55ee45e22f21aee5&vid=e30a0c577435467dc1a83c9b55ee45e22f21aee5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR