α'-Chloro-α,α,α-trifluoro-o-xylene ≥98.0% (by GC)

Supplier: TCI America

T2731-25G T2731-5G
TCT2731-25GEA 155.9 USD
TCT2731-25G TCT2731-5G
α'-Chloro-α,α,α-trifluoro-o-xylene ≥98.0% (by GC)
α'-Chloro-α,α,α-trifluoro-o-xylene

CAS Number: 21742-00-7
MDL Number: MFCD00009920
Molecular Formula: C8H6ClF3
Molecular Weight: 194.58
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Flash Point (°C): 70
Specific Gravity (20/20): 1.34


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Chloromethyl)benzotrifluoride, alpha-Chloro-alpha',alpha',alpha'-trifluoro-o-xlylene

Formula: C₈H₆ClF₃
Boiling Pt: 110 °C
Density: 1.34 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00009920
CAS Number: 21742-00-7
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d116c759e4eafa29aab9da282c3ee71eb426781&vid=8d116c759e4eafa29aab9da282c3ee71eb426781&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR