α-Bromo-3-(trifluoromethoxy)toluene ≥97.0% (by GC)

Supplier: TCI America

T2730-1G T2730-5G
TCT2730-1GEA 56 USD
TCT2730-1G TCT2730-5G
α-Bromo-3-(trifluoromethoxy)toluene ≥97.0% (by GC)
α-Bromo-3-(trifluoromethoxy)toluene

CAS Number: 159689-88-0
MDL Number: MFCD00061271
Molecular Formula: C8H6BrF3O
Molecular Weight: 255.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 179
Flash Point (°C): 89
Specific Gravity (20/20): 1.58


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-3-(trifluoromethoxy)toluene

Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Boiling Pt: 179 °C
Density: 1.58 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00061271
CAS Number: 159689-88-0
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b1b54f196cb9cc2b72d72184b37b9550e57574a&vid=0b1b54f196cb9cc2b72d72184b37b9550e57574a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR