1,3,5-Trimethyl-1H-pyrazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3,5-Trimethyl-1H-pyrazole, 1,3,5-Trimethylpyrazole

T2724-25G T2724-5G
TCT2724-25GEA 77 USD
TCT2724-25G TCT2724-5G
1,3,5-Trimethyl-1H-pyrazole ≥98.0% (by GC)
1,3,5-Trimethyl-1H-pyrazole

CAS Number: 1072-91-9
MDL Number: MFCD00015536
Molecular Formula: C6H10N2
Molecular Weight: 110.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 170
Melting point (°C): 37
Flash Point (°C): 40
Lambda max.: 220 nm (MeOH)

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 170 °C
Melting Pt: 37 °C
Flash Pt: 40 °C
MDL Number: MFCD00015536
CAS Number: 1072-91-9
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e336510b711927a61fa5acf81fdc18130d84de0b&vid=e336510b711927a61fa5acf81fdc18130d84de0b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR