1,3,5-Tris(bromomethyl)benzene ≥98.0% (by GC)

Supplier: TCI America

T2719-1G T2719-5G
TCT2719-1GEA 48 USD
TCT2719-1G TCT2719-5G
1,3,5-Tris(bromomethyl)benzene ≥98.0% (by GC)
1,3,5-Tris(bromomethyl)benzene

CAS Number: 18226-42-1
MDL Number: MFCD00182500
Molecular Formula: C9H9Br3
Molecular Weight: 356.88
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 99


Product Categories: Materials Chemistry, Dendrimer Building Blocks, Bioscience, Antibody Drug Research
Other Product Names / Synonyms: alpha,alpha',alpha''-Tribromomesitylene

Formula: C₉H₉Br₃
MW: 356.88 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00182500
CAS Number: 18226-42-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=853f072c7445cf6a92405845570d2964e608d0f2&vid=853f072c7445cf6a92405845570d2964e608d0f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR