Trimethylsilyl trifluoroacetate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Trifluoroacetic Acid Trimethylsilyl Ester

T2629-25G T2629-5G
TCT2629-25GEA 175 USD
TCT2629-25G TCT2629-5G
Trimethylsilyl trifluoroacetate ≥95.0% (by GC)
Trimethylsilyl trifluoroacetate

CAS Number: 400-53-3
MDL Number: MFCD00000413
Molecular Formula: C5H9F3O2Si
Molecular Weight: 186.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 90
Flash Point (°C): 13
Specific Gravity (20/20): 1.07


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Trifluoroacetic Acid Trimethylsilyl Ester

Formula: C₅H₉F₃O₂Si
MW: 186.21 g/mol
Boiling Pt: 90 °C
Density: 1.07 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00000413
CAS Number: 400-53-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceeddf9c4e0a0d322e6cac2c320ee33d57f8b230&vid=ceeddf9c4e0a0d322e6cac2c320ee33d57f8b230&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR