3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-2,2-bis(trifluoromethyl)acetic Acid

T2587-1G T2587-5G
TCT2587-1GEA 92 USD
TCT2587-1G TCT2587-5G
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid ≥98.0% (by titrimetric analysis)
3,3,3-Trifluoro-2-hydroxy-2-(trifluoromethyl)propionic acid

CAS Number: 662-22-6
MDL Number: MFCD00190631
Molecular Formula: C4H2F6O3
Molecular Weight: 212.05
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 153
Melting point (°C): 84


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Hydroxy-2,2-bis(trifluoromethyl)acetic Acid

Formula: C₄H₂F₆O₃
MW: 212.05 g/mol
Boiling Pt: 153 °C
Melting Pt: 84 °C
MDL Number: MFCD00190631
CAS Number: 662-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=532ce62b0f325113c41735bfe9a787343388469a&vid=532ce62b0f325113c41735bfe9a787343388469a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR