(R)-(-)-Tetrahydrofurfurylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-(-)-2-(Aminomethyl)tetrahydrofuran, (R)-2-Tetrahydrofurfurylamine

T2551-1G T2551-200MG
TCT2551-1GEA 174 USD
TCT2551-1G TCT2551-200MG
(R)-(-)-Tetrahydrofurfurylamine ≥98.0% (by GC, titration analysis)
(R)-(-)-Tetrahydrofurfurylamine

CAS Number: 7202-43-9
MDL Number: MFCD00192476
Molecular Formula: C5H11NO
Molecular Weight: 101.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Flash Point (°C): 46
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: -15 deg (C=2, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-2-(Aminomethyl)tetrahydrofuran

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 60 °C
Density: 0.98 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00192476
CAS Number: 7202-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32ea1cd9780111288596a813791a547089cf3329&vid=32ea1cd9780111288596a813791a547089cf3329&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR