2,2,2-Trifluoro-N-phenylacetimidoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2547-25G
T2547-5G
TCT2547-25GEA
1332
USD
TCT2547-25G
TCT2547-5G
2,2,2-Trifluoro-N-phenylacetimidoyl chloride ≥98.0% (by GC, titration analysis)
2,2,2-Trifluoro-N-phenylacetimidoyl chloride
CAS Number: 61881-19-4
Molecular Formula: C8H5ClF3N
Molecular Weight: 207.58
Purity/Analysis Method:
Formula:
C₈H₅ClF₃N MW: 207.58 g/mol Boiling Pt: 53 °C Density: 1.31 (20 °C) |
MDL Number:
MFCD11042687 CAS Number: 61881-19-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...