4-(Trifluoromethoxy)phenyl isocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Isocyanato-4-(trifluoromethoxy)benzene

T2487-25G T2487-5G
TCT2487-25GEA 76.3 USD
TCT2487-25G TCT2487-5G
4-(Trifluoromethoxy)phenyl isocyanate ≥98.0% (by GC)
4-(Trifluoromethoxy)phenyl isocyanate

CAS Number: 35037-73-1
MDL Number: MFCD00035702
Molecular Formula: C8H4F3NO2
Molecular Weight: 203.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 78
Flash Point (°C): 69
Specific Gravity (20/20): 1.36


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isocyanic Acid 4-(Trifluoromethoxy)phenyl Ester

Formula: C₈H₄F₃NO₂
MW: 203.12 g/mol
Boiling Pt: 78 °C
Density: 1.36 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00035702
CAS Number: 35037-73-1
UN: 2206
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3cb413cf82fd67c53af4cce049689320f0d3b0f&vid=e3cb413cf82fd67c53af4cce049689320f0d3b0f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR