1-(Trimethylsilyl)-2-naphthyl trifluoromethanesulfonate ≥96.0%
Supplier: TCI America
Synonyms:
Trifluoromethanesulphonic acid 1-(Trimethylsilyl)-2-naphthyl Ester, 1-(Trimethylsilyl)-2-naphthyl Triflate
CAS Number: 252054-88-9
Molecular Formula: C14H15F3O3SSi
Molecular Weight: 348.41
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.28
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Benzyne Precursors (Multi-component Reaction), Silicon Compounds (for Synthesis), Naphthalyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 1-(Trimethylsilyl)-2-naphthyl Triflate, Trifluoromethanesulfonic Acid 1-(Trimethylsilyl)-2-naphthyl Ester
Formula:
C₁₄H₁₅F₃O₃SSi Density: 1.28 (20 °C) |
CAS Number:
252054-88-9 |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...