3,4,5-Trifluorobenzyl bromide ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
5-(Bromomethyl)-1,2,3-trifluorobenzene, α-Bromo-3,4,5-trifluorotoluene
T2464-1G
T2464-5G
TCT2464-1GEA
22
USD
TCT2464-1G
TCT2464-5G
3,4,5-Trifluorobenzyl bromide ≥97.0% (by GC)
3,4,5-Trifluorobenzyl bromide
CAS Number: 220141-72-0
MDL Number: MFCD00083528
Molecular Formula: C7H4BrF3
Molecular Weight: 225.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 46
Flash Point (°C): 65
Specific Gravity (20/20): 1.71
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(Bromomethyl)-1,2,3-trifluorobenzene
Formula:
C₇H₄BrF₃ MW: 225.01 g/mol Boiling Pt: 46 °C Density: 1.71 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00083528 CAS Number: 220141-72-0 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...