2,3,4-Trifluorobenzyl bromide ≥98.0%

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-2,3,4-trifluorobenzene, α-Bromo-2,3,4-trifluorotoluene

T2462-1G T2462-5G
TCT2462-1GEA 39 USD
TCT2462-1G TCT2462-5G
2,3,4-Trifluorobenzyl bromide ≥98.0%
2,3,4-Trifluorobenzyl bromide

CAS Number: 157911-55-2
MDL Number: MFCD00061233
Molecular Formula: C7H4BrF3
Molecular Weight: 225.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Flash Point (°C): 91
Specific Gravity (20/20): 1.72


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-2,3,4-trifluorobenzene

Formula: C₇H₄BrF₃
Density: 1.72 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00061233
CAS Number: 157911-55-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=819c7fc837c503aaf253445a72866992522ab0e4&vid=819c7fc837c503aaf253445a72866992522ab0e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR