2-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: α,α,α-Trifluoro-o-tolyl isothiocyanate

T2459-1G T2459-5G
TCT2459-1GEA 23 USD
TCT2459-1G TCT2459-5G
2-(Trifluoromethyl)phenyl isothiocyanate ≥98.0% (by GC)
2-(Trifluoromethyl)phenyl isothiocyanate

CAS Number: 1743-86-8
MDL Number: MFCD00039644
Molecular Formula: C8H4F3NS
Molecular Weight: 203.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 216
Flash Point (°C): 105
Specific Gravity (20/20): 1.35


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isothiocyanic Acid 2-(Trifluoromethyl)phenyl Ester, alpha,alpha,alpha-Trifluoro-o-tolyl Isothiocyanate

Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 216 °C
Density: 1.35 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00039644
CAS Number: 1743-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06356faaccb92ecaf18a41b84daa46fb6fe57a5e&vid=06356faaccb92ecaf18a41b84daa46fb6fe57a5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR