4-(Trifluoromethoxy)toluene ≥98.0% (by GC)
Supplier: TCI America
T2454-1G
T2454-5G
TCT2454-1GEA
75.08
USD
TCT2454-1G
TCT2454-5G
4-(Trifluoromethoxy)toluene ≥98.0% (by GC)
4-(Trifluoromethoxy)toluene
CAS Number: 706-27-4
MDL Number: MFCD00042413
Molecular Formula: C8H7F3O
Molecular Weight: 176.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 135
Flash Point (°C): 31
Specific Gravity (20/20): 1.18
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Methyl-4-(trifluoromethoxy)benzene
Formula:
C₈H₇F₃O Boiling Pt: 135 °C Density: 1.18 (20 °C) Flash Pt: 31 °C |
MDL Number:
MFCD00042413 CAS Number: 706-27-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...