4-(Trifluoromethylthio)benzyl bromide ≥97.0% (by GC)

Supplier: TCI America

Synonyms: (4-(Bromomethyl)phenyl)(trifluoromethyl)sulphane

T2391-1G T2391-5G
TCT2391-1GEA 46 USD
TCT2391-1G TCT2391-5G
4-(Trifluoromethylthio)benzyl bromide ≥97.0% (by GC)
4-(Trifluoromethylthio)benzyl bromide

CAS Number: 21101-63-3
MDL Number: MFCD00129189
Molecular Formula: C8H6BrF3S
Molecular Weight: 271.10
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 118
Melting point (°C): 55
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-4-(trifluoromethylthio)toluene

Formula: C₈H₆BrF₃S
MW: 271.10 g/mol
Boiling Pt: 118 °C
Melting Pt: 55 °C
Flash Pt: 110 °C
MDL Number: MFCD00129189
CAS Number: 21101-63-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=083e247fff92445434dd325c1d2395988b3aa0dd&vid=083e247fff92445434dd325c1d2395988b3aa0dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR