Triethylsilylacetylene ≥96.0% (by GC)

Supplier: TCI America

T2387-1G T2387-5G
TCT2387-1GEA 23 USD
TCT2387-1G TCT2387-5G
Triethylsilylacetylene ≥96.0% (by GC)
Triethylsilylacetylene

CAS Number: 1777-03-3
MDL Number: MFCD00075373
Molecular Formula: C8H16Si
Molecular Weight: 140.30
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 136
Flash Point (°C): 17
Specific Gravity (20/20): 0.78


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Click Chemistry
Other Product Names / Synonyms: Ethynyltriethylsilane, Triethylethynylsilane

Formula: C₈H₁₆Si
MW: 140.3 g/mol
Boiling Pt: 136 °C
Density: 0.78 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00075373
CAS Number: 1777-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=146f3a8d2bfdaecc3b586722af8d9b817f3e6b25&vid=146f3a8d2bfdaecc3b586722af8d9b817f3e6b25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR