2,4,5-Trifluorobenzyl bromide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene

T2386-1G T2386-5G
TCT2386-1GEA 32 USD
TCT2386-1G TCT2386-5G
2,4,5-Trifluorobenzyl bromide ≥98.0% (by GC, titration analysis)
2,4,5-Trifluorobenzyl bromide

CAS Number: 157911-56-3
MDL Number: MFCD00061209
Molecular Formula: C7H4BrF3
Molecular Weight: 225.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.71


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene

Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Density: 1.71 (20 °C)
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955c2b8d1a3fafc7a43a9724214938771fa94319&vid=955c2b8d1a3fafc7a43a9724214938771fa94319&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR