3-(Trifluoromethoxy)benzonitrile ≥98.0%

Supplier: TCI America

T2372-1G T2372-5G
TCT2372-1GEA 38 USD
TCT2372-1G TCT2372-5G
3-(Trifluoromethoxy)benzonitrile ≥98.0%
3-(Trifluoromethoxy)benzonitrile

CAS Number: 52771-22-9
MDL Number: MFCD00040959
Molecular Formula: C8H4F3NO
Molecular Weight: 187.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.31

Formula: C₈H₄F₃NO
Density: 1.31 (20 °C)
MDL Number: MFCD00040959
CAS Number: 52771-22-9
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49b658bd8e70ad8180da3b133ce3e007bed65aa6&vid=49b658bd8e70ad8180da3b133ce3e007bed65aa6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR