3,4,5-Trifluorobenzonitrile ≥98.0% (by GC, total nitrogen)

Supplier: TCI America

T2370-1G T2370-5G
TCT2370-1GEA 39 USD
TCT2370-1G TCT2370-5G
3,4,5-Trifluorobenzonitrile ≥98.0% (by GC, total nitrogen)
3,4,5-Trifluorobenzonitrile

CAS Number: 134227-45-5
MDL Number: MFCD00074948
Molecular Formula: C7H2F3N
Molecular Weight: 157.10
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Color: White
Boiling point (°C): 69
Melting point (°C): 47
Flash Point (°C): 63

Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 69 °C
Melting Pt: 47 °C
Flash Pt: 63 °C
MDL Number: MFCD00074948
CAS Number: 134227-45-5
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6018752c6f900ec809c871f7971bb1e8624fd578&vid=6018752c6f900ec809c871f7971bb1e8624fd578&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR