1,2,3-Trifluorobenzene ≥98.0%

Supplier: TCI America

T2356-1G T2356-5G
TCT2356-1GEA 22 USD
TCT2356-1G TCT2356-5G
1,2,3-Trifluorobenzene ≥98.0%
1,2,3-Trifluorobenzene

CAS Number: 1489-53-8
MDL Number: MFCD00012013
Molecular Formula: C6H3F3
Molecular Weight: 132.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 95
Flash Point (°C): -4
Specific Gravity (20/20): 1.30

Formula: C₆H₃F₃
Boiling Pt: 95 °C
Density: 1.30 (20 °C)
Flash Pt: –4 °C
MDL Number: MFCD00012013
CAS Number: 1489-53-8
UN: 2387
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04c5885b35ea666442620e0c6b87c8b2c312d5a0&vid=6bfe8188decb168d48d335326f0bec32491da504&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR