3-(Trifluoromethoxy)benzoyl chloride ≥98.0% (by GC)

Supplier: TCI America

T2334-1G T2334-5G
TCT2334-1GEA 47 USD
TCT2334-1G TCT2334-5G
3-(Trifluoromethoxy)benzoyl chloride ≥98.0% (by GC)
3-(Trifluoromethoxy)benzoyl chloride

CAS Number: 86270-03-3
MDL Number: MFCD00061268
Molecular Formula: C8H4ClF3O2
Molecular Weight: 224.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.43

Formula: C₈H₄ClF₃O₂
Density: 1.43 (20 °C)
MDL Number: MFCD00061268
CAS Number: 86270-03-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e471e784ec010dd6bf0304cdefcb09a10151072c&vid=e471e784ec010dd6bf0304cdefcb09a10151072c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR