2,3,5,6-Tetrachloropyridine ≥98.0% (by GC)
Supplier: TCI America
T2331-25G
T2331-5G
TCT2331-25GEA
205
USD
TCT2331-25G
TCT2331-5G
2,3,5,6-Tetrachloropyridine ≥98.0% (by GC)
2,3,5,6-Tetrachloropyridine
CAS Number: 2402-79-1
Molecular Formula: C5HCl4N
Molecular Weight: 216.87
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 251
Melting point (°C): 93
Flash Point (°C): 188
Formula:
C₅HCl₄N MW: 216.88 g/mol Boiling Pt: 251 °C Melting Pt: 93 °C Flash Pt: 188 °C |
MDL Number:
MFCD00092428 CAS Number: 2402-79-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...