2,4,5-Trifluorobenzonitrile ≥98.0% (by GC)
Supplier: TCI America
T2326-25G
T2326-5G
TCT2326-25GEA
207.1
USD
TCT2326-25G
TCT2326-5G
2,4,5-Trifluorobenzonitrile ≥98.0% (by GC)
2,4,5-Trifluorobenzonitrile
CAS Number: 98349-22-5
MDL Number: MFCD00013289
Molecular Formula: C7H2F3N
Molecular Weight: 157.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 170
Flash Point (°C): 83
Specific Gravity (20/20): 1.38
Formula:
C₇H₂F₃N MW: 157.09 g/mol Boiling Pt: 170 °C Density: 1.38 (20 °C) Flash Pt: 83 °C |
MDL Number:
MFCD00013289 CAS Number: 98349-22-5 UN: 3276 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...