2,4,5-Trifluorobenzaldehyde ≥98.0% (by GC)

Supplier: TCI America

T2319-1G T2319-5G
TCT2319-1GEA 38 USD
TCT2319-1G TCT2319-5G
2,4,5-Trifluorobenzaldehyde ≥98.0% (by GC)
2,4,5-Trifluorobenzaldehyde

CAS Number: 165047-24-5
MDL Number: MFCD00061196
Molecular Formula: C7H3F3O
Molecular Weight: 160.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 168
Flash Point (°C): 59
Specific Gravity (20/20): 1.42

Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 168 °C
Density: 1.42 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00061196
CAS Number: 165047-24-5
UN: 1989
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc437366df19aae8f54199a250fff23d441ee30c&vid=bc437366df19aae8f54199a250fff23d441ee30c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR