2-(Trifluoromethyl)benzylamine ≥98.0%
Supplier: TCI America
Synonyms:
(2-(Trifluoromethyl)phenyl)methanamine
T2315-25G
T2315-5G
TCT2315-25GEA
257
USD
TCT2315-25G
TCT2315-5G
2-(Trifluoromethyl)benzylamine ≥98.0%
2-(Trifluoromethyl)benzylamine
CAS Number: 3048-01-9
MDL Number: MFCD00010297
Molecular Formula: C8H8F3N
Molecular Weight: 175.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 110
Flash Point (°C): 69
Specific Gravity (20/20): 1.26
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-(Aminomethyl)benzotrifluoride
Formula:
C₈H₈F₃N Boiling Pt: 110 °C Density: 1.26 (20 °C) Flash Pt: 69 °C |
MDL Number:
MFCD00010297 CAS Number: 3048-01-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...