α-Bromo-2-(trifluoromethoxy)toluene ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 198649-68-2
MDL Number: MFCD00236321
Molecular Formula: C8H6BrF3O
Molecular Weight: 255.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow
Specific Gravity (20/20): 1.59
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-2-(trifluoromethoxy)toluene
Formula:
C₈H₆BrF₃O Density: 1.59 (20 °C) |
MDL Number:
MFCD00236321 CAS Number: 198649-68-2 UN: 3265 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...