3,4,5-Trifluorophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2299-25G T2299-5G
TCT2299-25GEA 150 USD
TCT2299-25G TCT2299-5G
3,4,5-Trifluorophenol ≥98.0% (by GC, titration analysis)
3,4,5-Trifluorophenol

CAS Number: 99627-05-1
MDL Number: MFCD00042427
Molecular Formula: C6H3F3O
Molecular Weight: 148.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 177
Melting point (°C): 57

Formula: C₆H₃F₃O
MW: 148.08 g/mol
Boiling Pt: 177 °C
Melting Pt: 57 °C
MDL Number: MFCD00042427
CAS Number: 99627-05-1
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fbf12f3bac16a19a81af9fbe512cf07dc6a2404&vid=8a90a24b6bf84f76fe9e80970a6619bbfc61515e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR