4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane ≥95.0% (by GC) stabilized
Supplier: TCI America
Synonyms:
Pinacol vinylboronate, Vinylboronic Acid Pinacol Ester
T2297-1G
T2297-25G
T2297-5G
TCT2297-1GEA
54
USD
TCT2297-1G
TCT2297-25G
TCT2297-5G
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane ≥95.0% (by GC) stabilized
4,4,5,5-Tetramethyl-2-vinyl-1,3,2-dioxaborolane
(stabilized with Phenothiazine)
CAS Number: 75927-49-0
Molecular Formula: C8H15BO2
Molecular Weight: 154.02
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 40
Flash Point (°C): 34
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Vinylation, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: Vinylboronic Acid Pinacol Ester
Formula:
C₈H₁₅BO₂ MW: 154.02 g/mol Boiling Pt: 40 °C Density: 0.90 (20 °C) Flash Pt: 34 °C |
MDL Number:
MFCD00192492 CAS Number: 75927-49-0 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...