α'-Chloro-α,α,α-trifluoro-m-xylene ≥95.0% (by GC)
Supplier: TCI America
CAS Number: 705-29-3
Molecular Formula: C8H6ClF3
Molecular Weight: 194.58
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Flash Point (°C): 49
Specific Gravity (20/20): 1.32
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-(Chloromethyl)benzotrifluoride, alpha-Chloro-alpha',alpha',alpha'-trifluoro-m-xylene
Formula:
C₈H₆ClF₃ Boiling Pt: 70 °C Density: 1.32 (20 °C) Flash Pt: 49 °C |
MDL Number:
MFCD00000908 CAS Number: 705-29-3 UN: 3265 ADR: 8,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...