Trimethyl orthopropionate ≥97.0% (by GC)
Supplier: TCI America
T2288-25G
T2288-5G
TCT2288-25GEA
46
USD
TCT2288-25G
TCT2288-5G
Trimethyl orthopropionate ≥97.0% (by GC)
Trimethyl orthopropionate
CAS Number: 24823-81-2
MDL Number: MFCD00043305
Molecular Formula: C6H14O3
Molecular Weight: 134.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 128
Flash Point (°C): 19
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Orthopropionic Acid Trimethyl Ester, 1,1,1-Trimethoxypropane, Methyl Orthopropionate
Formula:
C₆H₁₄O₃ MW: 134.18 g/mol Boiling Pt: 128 °C Density: 0.95 (20 °C) Flash Pt: 19 °C |
MDL Number:
MFCD00043305 CAS Number: 24823-81-2 UN: 3272 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...