Trimethylamine ∼25% ∼3.2 mol/L in methanol

Supplier: TCI America

T2268-500ML T2268-100ML
TCT2268-500MLEA 100 USD
TCT2268-500ML TCT2268-100ML
Trimethylamine ∼25% ∼3.2 mol/L in methanol
Trimethylamine

CAS Number: 75-50-3
MDL Number: MFCD00008327
Molecular Formula: C3H9N
Molecular Weight: 59.11
Form: Clear Liquid
Color: Colorless
Storage Temperature: 0-10°C

Formula: C₃H₉N
MW: 59.11 g/mol
MDL Number: MFCD00008327
CAS Number: 75-50-3
UN: 1297
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab7ef2cc7e2b7c936ce95e216ff7a4425ec52147&vid=88c044e92baed75b10440243f55fdfa15a97e641&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR