2,4,5-Tribromoimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2,4,5-Tribromo-1H-imidazole

T2266-25G T2266-5G
TCT2266-25GEA 111 USD
TCT2266-25G TCT2266-5G
2,4,5-Tribromoimidazole ≥98.0% (by HPLC, titration analysis)
2,4,5-Tribromoimidazole

CAS Number: 2034-22-2
Molecular Formula: C3HBr3N2
Molecular Weight: 304.77
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 212

Formula: C₃HBr₃N₂
MW: 304.77 g/mol
Melting Pt: 212 °C
MDL Number: MFCD00005184
CAS Number: 2034-22-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR