2,2,3,3-Tetrafluoropropyl acrylate ≥98.0% stabilized

Supplier: TCI America

Synonyms: Acrylic Acid 2,2,3,3-Tetrafluoropropyl Ester

T2240-25G T2240-5G
TCT2240-25GEA 111 USD
TCT2240-25G TCT2240-5G
2,2,3,3-Tetrafluoropropyl acrylate ≥98.0% stabilized
2,2,3,3-Tetrafluoropropyl acrylate

(stabilized with MEHQ)
CAS Number: 7383-71-3
Molecular Formula: C6H6F4O2
Molecular Weight: 186.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 75
Flash Point (°C): 46
Specific Gravity (20/20): 1.31
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 2,2,3,3-Tetrafluoropropyl Ester

Formula: C₆H₆F₄O₂
MW: 186.11 g/mol
Boiling Pt: 75 °C
Density: 1.31 (20 °C)
Flash Pt: 46 °C
CAS Number: 7383-71-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f291a9fbc0011a894a7896616a67d6ffec3fc9&vid=31f291a9fbc0011a894a7896616a67d6ffec3fc9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR