N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate ≥98.0%

Supplier: TCI America

Synonyms: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate

T2224-1G T2224-5G
TCT2224-1GEA 38 USD
TCT2224-1G TCT2224-5G
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate ≥98.0%
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate

CAS Number: 105832-38-0
Molecular Formula: C9H16BF4N3O3
Molecular Weight: 301.05
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 195
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Peptide Synthesis
Other Product Names / Synonyms: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU

Formula: C₉H₁₆BF₄N₃O₃
Melting Pt: 195 °C
MDL Number: MFCD00077875
CAS Number: 105832-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb6c67e8aa7a3aa3fc8db55f2471d9160d5b939e&vid=cb6c67e8aa7a3aa3fc8db55f2471d9160d5b939e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR