1-Bromo-2-(trifluoromethoxy)benzene ≥97.0% (by GC)

Supplier: TCI America

T2147-25G T2147-5G
TCT2147-25GEA 34 USD
TCT2147-25G TCT2147-5G
1-Bromo-2-(trifluoromethoxy)benzene ≥97.0% (by GC)
1-Bromo-2-(trifluoromethoxy)benzene

CAS Number: 64115-88-4
Molecular Formula: C7H4BrF3O
Molecular Weight: 241.01
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 160
Flash Point (°C): 60
Specific Gravity (20/20): 1.64

Formula: C₇H₄BrF₃O
Boiling Pt: 160 °C
Density: 1.64 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00040942
CAS Number: 64115-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b997d19c08f184182024f99042b71c269394c1b5&vid=b997d19c08f184182024f99042b71c269394c1b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR