2-(Trifluoromethoxy)aniline ≥98.0%

Supplier: TCI America

T2145-25G T2145-5G
TCT2145-25GEA 56 USD
TCT2145-25G TCT2145-5G
2-(Trifluoromethoxy)aniline ≥98.0%
2-(Trifluoromethoxy)aniline

CAS Number: 1535-75-7
Molecular Formula: C7H6F3NO
Molecular Weight: 177.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 63
Flash Point (°C): 54
Specific Gravity (20/20): 1.32


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-o-anisidine

Formula: C₇H₆F₃NO
Boiling Pt: 63 °C
Density: 1.32 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00035959
CAS Number: 1535-75-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37b5996cf78f289767ed7ab80b89f768d40a830e&vid=37b5996cf78f289767ed7ab80b89f768d40a830e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR