Sulfur trioxide triethylamine complex ≥95.0% (by total nitrogen basis)
Supplier: TCI America
CAS Number: 761-01-3
Molecular Formula: C6H15NO3S
Molecular Weight: 181.25
Purity/Analysis Method: >95.0% (N)
Form: Crystal
Color: White
Melting point (°C): 93
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis)
Other Product Names / Synonyms: Triethylamine - Sulfur Trioxide Complex
Formula:
C₆H₁₅NO₃S Melting Pt: 93 °C |
CAS Number:
761-01-3 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >95.0% (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...