2-Thiophenecarbonitrile ≥98.0% (by GC)
Supplier: TCI America
CAS Number: 1003-31-2
MDL Number: MFCD00005416
Molecular Formula: C5H3NS
Molecular Weight: 109.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 192
Flash Point (°C): 53
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: 2-Thiophenecarbonitrile
Formula:
C₅H₃NS Boiling Pt: 192 °C Density: 1.18 (20 °C) Flash Pt: 53 °C |
MDL Number:
MFCD00005416 CAS Number: 1003-31-2 UN: 3276 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...