2-(Trimethylsilyl)phenyl trifluoromethanesulfonate ≥95.0%

Supplier: TCI America

Synonyms: Trifluoromethanesulphonic acid 2-(Trimethylsilyl)phenyl Ester, 2-(Trimethylsilyl)phenyl Triflate

T2089-1G T2089-5G
TCT2089-1GEA 63 USD
TCT2089-1G TCT2089-5G
2-(Trimethylsilyl)phenyl trifluoromethanesulfonate ≥95.0%
2-(Trimethylsilyl)phenyl trifluoromethanesulfonate

CAS Number: 88284-48-4
MDL Number: MFCD00799598
Molecular Formula: C10H13F3O3SSi
Molecular Weight: 298.35
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 70
Flash Point (°C): 96
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Benzyne Precursors (Multi-component Reaction), Silicon Compounds (for Synthesis), Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 2-(Trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-(Trimethylsilyl)phenyl Ester

Formula: C₁₀H₁₃F₃O₃SSi
MW: 298.35 g/mol
Boiling Pt: 70 °C
Density: 1.23 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00799598
CAS Number: 88284-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f85b237c6e4563f9560bb1dfc06d44b293f9b099&vid=f85b237c6e4563f9560bb1dfc06d44b293f9b099&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR