2,4,6-Trichlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2035-25G T2035-5G
TCT2035-25GEA 345 USD
TCT2035-25G TCT2035-5G
2,4,6-Trichlorobenzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
2,4,6-Trichlorobenzenesulphonyl chloride

CAS Number: 51527-73-2
MDL Number: MFCD00052973
Molecular Formula: C6H2Cl4O2S
Molecular Weight: 279.94
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 49

Formula: C₆H₂Cl₄O₂S
Melting Pt: 49 °C
MDL Number: MFCD00052973
CAS Number: 51527-73-2
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ab7a8a9a3f8a8e7e348edbc61cb4e1bc4b0e36d&vid=9ab7a8a9a3f8a8e7e348edbc61cb4e1bc4b0e36d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR