exo-Tetrahydrodicyclopentadiene ≥94.0% (by GC)
Supplier: TCI America
Synonyms:
exo-Tricyclo[5.2.1.0(2,6)]decane, (3aR,4S,7R,7aS)-rel-Octahydro-1H-4,7-methanoindene
T1994-25G
T1994-5G
TCT1994-25GEA
532
USD
TCT1994-25G
TCT1994-5G
exo-Tetrahydrodicyclopentadiene ≥94.0% (by GC)
exo-Tetrahydrodicyclopentadiene
CAS Number: 2825-82-3
MDL Number: MFCD01714931
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 55
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: exo-Tricyclo[5.2.1.0(2,6)]decane
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Density: 0.94 (20 °C) Flash Pt: 55 °C |
MDL Number:
MFCD01714931 CAS Number: 2825-82-3 |
Specification Test Results
Purity/Analysis Method | >94.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...