2,2,4-Trimethyl-2-pentanol ≥95.0% (by GC)

Supplier: TCI America

T1962-1G T1962-5G
TCT1962-1GEA 102 USD
TCT1962-1G TCT1962-5G
2,2,4-Trimethyl-2-pentanol ≥95.0% (by GC)
2,2,4-Trimethyl-2-pentanol

CAS Number: 690-37-9
MDL Number: MFCD00101611
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: 2-Hydroxy-2,4,4-trimethylpentane

Formula: C₈H₁₈O
Density: 0.82 (20 °C)
MDL Number: MFCD00101611
CAS Number: 690-37-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6420a30ed943eeb6d180d84f3b8c7964578c8551&vid=6420a30ed943eeb6d180d84f3b8c7964578c8551&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR