(Triphenylphosphoranylidene)acetonitrile ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
2-(Triphenylphosphoranylidene)acetonitrile, (Cyanomethylene)triphenylphosphorane
T1958-25G
T1958-5G
TCT1958-25GEA
672.61
USD
TCT1958-25G
TCT1958-5G
(Triphenylphosphoranylidene)acetonitrile ≥98.0% (by HPLC, total nitrogen)
(Triphenylphosphoranylidene)acetonitrile
CAS Number: 16640-68-9
MDL Number: MFCD00567633
Molecular Formula: C20H16NP
Molecular Weight: 301.33
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 193
Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (Cyanomethylene)triphenylphosphorane
Formula:
C₂₀H₁₆NP MW: 301.33 g/mol Melting Pt: 193 °C |
MDL Number:
MFCD00567633 CAS Number: 16640-68-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...