2,3,5,6-Tetrafluoro-p-xylene ≥98.0% (by GC)

Supplier: TCI America

T1947-1G T1947-5G
TCT1947-1GEA 68 USD
TCT1947-1G TCT1947-5G
2,3,5,6-Tetrafluoro-p-xylene ≥98.0% (by GC)
2,3,5,6-Tetrafluoro-p-xylene

CAS Number: 703-87-7
MDL Number: MFCD00012231
Molecular Formula: C8H6F4
Molecular Weight: 178.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 35
Flash Point (°C): 47


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3,5,6-Tetrafluoro-1,4-dimethylbenzene

Formula: C₈H₆F₄
Melting Pt: 35 °C
Flash Pt: 47 °C
MDL Number: MFCD00012231
CAS Number: 703-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db764d0b3b6b123929a882cfc49296923cdf7b84&vid=db764d0b3b6b123929a882cfc49296923cdf7b84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR