Yttrium(III) trifluoromethanesulphonate ≥98.0%

Supplier: TCI America

Synonyms: Yttrium(III) trifluoromethanesulphonate, Yttrium triflate

T1921-25G T1921-5G
TCT1921-25GEA 235 USD
TCT1921-25G TCT1921-5G
Yttrium(III) trifluoromethanesulphonate ≥98.0%
Yttrium(III) trifluoromethanesulphonate

CAS Number: 52093-30-8
MDL Number: MFCD00209623
Molecular Formula: CHF3O3S
Molecular Weight: 536.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Stable Lewis Acids in Aqueous Media
Other Product Names / Synonyms: Yttrium(III) Triflate, Trifluoromethanesulfonic Acid Yttrium(III) Salt

Formula: C₃F₉O₉S₃Y
MW: 536.12 g/mol
MDL Number: MFCD00209623
CAS Number: 52093-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ca77a5f3979b68c1fdbc1eaa8838c1f3888b56c&vid=8ca77a5f3979b68c1fdbc1eaa8838c1f3888b56c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR