3-(Trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Trifluoromethyl)benzene-1-sulphonyl chloride

T1910-25G T1910-5G
TCT1910-25GEA 174 USD
TCT1910-25G TCT1910-5G
3-(Trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3-(Trifluoromethyl)benzenesulphonyl chloride

CAS Number: 777-44-6
MDL Number: MFCD00014724
Molecular Formula: C7H4ClF3O2S
Molecular Weight: 244.61
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 110
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: alpha,alpha,alpha-Trifluorotoluene-3-sulfonyl Chloride

Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Density: 1.53 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00014724
CAS Number: 777-44-6
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3292779db8c63485b9d4d03dffcaf94202b16ab&vid=b3292779db8c63485b9d4d03dffcaf94202b16ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR