Tetrabutylammonium difluorotriphenylsilicate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

T1909-25G T1909-5G
TCT1909-25GEA 322 USD
TCT1909-25G TCT1909-5G
Tetrabutylammonium difluorotriphenylsilicate ≥97.0% (by titrimetric analysis)
Tetrabutylammonium difluorotriphenylsilicate

CAS Number: 163931-61-1
MDL Number: MFCD00274218
Molecular Formula: C34H51F2NSi
Molecular Weight: 539.87
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 156


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents
Other Product Names / Synonyms: TBAT

Formula: C₃₄H₅₁F₂NSi
Melting Pt: 156 °C
MDL Number: MFCD00274218
CAS Number: 163931-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b547dc0bc87b5ac47b8e3039d787aee321ec357d&vid=b547dc0bc87b5ac47b8e3039d787aee321ec357d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR