3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3,3,3-Trifluoro-2-hydroxyisobutyric Acid, 3,3,3-Trifluoro-2-hydroxy-2-methylpropanoic acid, 2-Hydroxy-2-(trifluoromethyl)propionic Acid
T1902-1G
T1902-5G
TCT1902-1GEA
74.3
USD
TCT1902-1G
TCT1902-5G
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid ≥97.0% (by titrimetric analysis)
3,3,3-Trifluoro-2-hydroxy-2-methylpropionic acid
CAS Number: 374-35-6
MDL Number: MFCD00190646
Molecular Formula: C4H5F3O3
Molecular Weight: 158.08
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Boiling point (°C): 189
Melting point (°C): 88
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Hydroxy-2-(trifluoromethyl)propionic Acid, 3,3,3-Trifluoro-2-hydroxyisobutyric Acid
Formula:
C₄H₅F₃O₃ MW: 158.08 g/mol Boiling Pt: 189 °C Melting Pt: 88 °C |
MDL Number:
MFCD00190646 CAS Number: 374-35-6 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...