1,4,7-Triazacyclononane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,4,7-Triazonane, TACN

T1878-1G T1878-200MG T1878-5G
TCT1878-1GEA 162.2 USD
TCT1878-1G TCT1878-200MG TCT1878-5G
1,4,7-Triazacyclononane ≥98.0% (by GC)
1,4,7-Triazacyclononane

CAS Number: 4730-54-5
Molecular Formula: C6H15N3
Molecular Weight: 129.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 113
Melting point (°C): 45
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Precise Polymerization Reagents, Macrocycles for Host-Guest Chemistry, Bioscience, Polyamines
Other Product Names / Synonyms: Octahydro-1H-1,4,7-triazonine

Formula: C₆H₁₅N₃
MW: 129.2 g/mol
Boiling Pt: 113 °C
Melting Pt: 45 °C
Flash Pt: 110 °C
MDL Number: MFCD00012358
CAS Number: 4730-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f18bf6fa8f0af36b39425415d8d6c9d6e1e4b5e&vid=8f18bf6fa8f0af36b39425415d8d6c9d6e1e4b5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR